Product Image
Invitrogen™
BLOCK-iT™ Lentiviral RNAi Expression System
The pLenti6/BLOCK-iT™-DEST expression vector provided in the BLOCK-iT™ Lentiviral RNAi Expression System can be used to efficiently introduce and stablyRead more
Have Questions?
Catalog NumberQuantity
K49440020 reactions
Catalog number K494400
Price (USD)
3,415.00
Each
Quantity:
20 reactions
Price (USD)
3,415.00
Each
The pLenti6/BLOCK-iT™-DEST expression vector provided in the BLOCK-iT™ Lentiviral RNAi Expression System can be used to efficiently introduce and stably express short hairpin RNA (shRNA) in vivo from a lentiviral vector. A novel cloning process places an ∼50-bp DNA oligonucleotide immediately following a U6 pol III promoter into the BLOCK-iT™ U6 entry vector. The oligonucleotide is designed to express RNA that forms a stem-loop structure containing the sense and antisense regions of your target gene of interest. This shRNA is then recombined into the pLenti6/BLOCK-iT™-DEST vector. After viral production and transduction, the shRNA driven by the U6 promoter becomes stably integrated as an RNAi cassette. The shRNA generated avoids the hosts defense mechanism and will be effective at producing the RNAi gene knockdown response (Figure 1).
The pLenti6/BLOCK-iT™-DEST vector (Figure 2) offers:
• attR sites for efficient recombination with the attL-flanked U6 Gateway™ entry vector containing the RNAi cassette
• All of the required components for efficient lentiviral packaging and delivery of the shRNA of interest
• Blasticidin selection marker for fast, efficient selection of stable cell lines expressing the shRNA Using the BLOCK-iT Lentiviral RNAi Expression System, long-term analysis of gene blocking in both dividing and non-dividing mammalian cell types and animal models can be achieved.
The BLOCK-iT™ RNAi U6 Entry Vector Kit allows streamlined cloning of shRNA target sequences for testing in transient experiments. Selected RNAi expression cassettes are quickly and efficiently recombined from the BLOCK-iT™ RNAi U6 entry vector into the pLenti6/BLOCK-iT™-DEST vector via a standard Gateway™ LR recombination reaction (Figure 3).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodGateway™
Constitutive or Inducible SystemConstitutive
Delivery TypeLentiviral
Product LineBLOCK-iT, Gateway
Product TypeRNAi Expression Vector Kit
Quantity20 reactions
RNAi TypeshRNA
Selection Agent (Eukaryotic)Blasticidin
VectorpLenti
PromoterU6
Unit SizeEach
Contents & Storage
The BLOCK-iT™ Lentiviral RNAi Gateway™ Vector Kit contains the pLenti6/BLOCK-iT™-DEST vector, pLenti6-GW/U6-laminshRNA control vector, and One Shot¤ Stbl3 Competent Cells. Vectors are supplied supercoiled and lyophilized. Store at -20°C once resuspended. Store Competent Cells at -80°C. The BLOCK-iT™ Lentiviral RNAi Expression System contains the BLOCK-iT™ Lentiviral RNAi Gateway™ Vector Kit, BLOCK-iT™ U6 Entry Vector Kit, ViraPower™ Bsd Support Kit, 293FT Cell Line, and LR Clonase™ enzyme mix. Store components as indicated in the respective kits. Store the LR Clonase™ mix at -80°C. Guaranteed stable for 6 months when properly stored.
Frequently asked questions (FAQs)
Can I use any Gateway entry vector to generate entry clones for use in RNAi applications?
No, you should use an entry vector that contains the elements necessary for RNA Polymerase III-dependent expression of your shRNA (i.e., Pol III promoter and terminator).
What is a dose response curve or kill curve? And can you outline the steps involved?
A dose response curve or kill curve is a simple method for determining the optimal antibiotic concentration to use when establishing a stable cell line. Untransfected cells are grown in a medium containing antibiotic at varying concentrations in order to determine the lowest amount of antibiotic needed to achieve complete cell death. The basic steps for performing a dose response curve or kill curve are as follows:
- Plate untransfected cells at 25% confluence, and grow them in a medium containing increasing concentrations of the antibiotic. For some antibiotics, you will need to calculate the amount of active drug to control for lot variation.
- Replenish the selective medium every 3-4 days. After 10-12 days, examine the dishes for viable cells. The cells may divide once or twice in the selective medium before cell death begins to occur.
- Look for the minimum concentration of antibiotic that resulted in complete cell death. This is the optimal antibiotic concentration to use for stable selection.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
Can I create stable cell lines using pENTR/U6 entry vector or the pENTR/H1/TO vector?
Unfortunately, the pENTR/U6 vector does not contain a selection marker; therefore, only transient RNAi analysis may be performed. If you wish to generate stable cell lines, perform an LR reaction into an appropriate Gateway destination vector to generate expression clones.
The pENTR/H1/TO vector contains the Zeocin resistance gene to facilitate generation of cell lines that inducbily express the shRNA of interest. Perform a kill curve to determine the minimum concentration of Zeocin that is required to kill your untransfected mammalian cell line. Please note that Zeocin-sensitive cells do not round up and detach from the plate, but rather may increase in size, show abnormal cell shape, display presence of large empty vesicles in the cytoplasm, or show breakdown of plasma/nuclear membranes.
Find additional tips, troubleshooting help, and resources within our RNAi Support Center.
What loop sequence should I use when designing my shRNA for cloning? Do you have any guidelines I should follow?
You can use a loop sequence of any length ranging from 4 to 11 nucleotides, although short loops (i.e., 4-7 nucleotides) are generally preferred. Avoid using a loop sequence containing thymidines (Ts), as they may cause early termination. This is particularly true if the target sequence itself ends in one or more T nucleotides. Here are some loop sequences we recommend:
- 5' - CGAA - 3'
- 5' - AACG - 3'
- 5' - GAGA - 3'
What considerations regarding transcription initiation should I take when designing my shRNA for cloning?
Transcription of the shRNA initiates at the first base following the end of the U6 promoter sequence. In the top-strand oligo, the transcription initiation site corresponds to the first nucleotide following the 4 bp CACC sequence added to permit directional cloning. We recommend initiating the shRNA sequence at a guanosine (G) because transcription of the native U6 snRNA initiates at a G. Note the following:
- If G is part of the target sequence, then incorporate the G into the stem sequence in the top-strand oligo and add a complementary C to the 3' end of the top-strand oligo.
- If G is not the first base of the target sequence, we recommend adding a G to the 5' end of the top-strand oligo directly following the CACC overhang sequence. In this case, do not add the complementary C to the 3' end of the top-strand oligo. Note: We have found that adding the complementary C in this situation can result in reduced activity of the shRNA. Alternative, if use of a G to initiate transcription is not desired, use an adenosine (A) rather than C or T. Note, however, that use of any nucleotide other than G may affect initiation efficiency and position.