Search
探索各種配件以提升您的微量盤分析儀的儀器性能,包括驗證微量盤、過濾片以及 µDrop 與 µDrop Duo 微量盤。
Using the right microplate is critical for the success of plate-based assays. Plastics used in the production of certain plates may have autofluorescence producing false positive results. For assays with live cells, coatings or special surfaces may be necessary to promote attachment and cell health. Thermo Fisher Scientific offers a range of microplates to support your assays.
Options include:
| Thermo Scientific Nunc cell culture plates | Nunc black and white polystyrene plates | Nunc Edge plates | Microplates for fluorescence-based assays |
| Choose from a wide selection of surface modifications and formats for a variety of 2D and 3D cell-based assays. | Get optimal performance with minimal background and crosstalk between wells for maximal signal detection. | Minimize evaporation concerns for live cell assays with long incubations. | Minimize well-to-well crosstalk and autofluorescence with these black-walled, clear-bottom 96- and 384-well microplates for fluorescence-based applications. |
Thermo Scientific μDrop and μDrop Duo Plates are excellent tools for quick and easy photometric measurement of DNA, RNA, and protein concentration in low sample volumes (2–10 μL), as well as spectral scanning. The low volume area is designed for use with an 8-channel pipette for fast and accurate multiple sample pipetting. The plates also contain a separate area for cuvette measurement. µDrop Plates are compatible with Thermo Scientific Varioskan LUX, Varioskan ALF, and Multiskan SkyHigh microplate readers. Our user-friendly SkanIt Software offers ready-made protocols for quick sample measurement and analysis using the μDrop and μDrop Duo Plates.
Thermo Scientific 微量盤分析儀驗證微量盤為一專用的性能驗證系統,可用於驗證並記錄 Thermo Scientific Multiskan FC、Multiskan SkyHigh、Luminoskan、Fluoroskan FL 以及 Varioskan LUX 的重要性能參數。
產品編號 |
介紹 |
產品包含 |
Spectrophotometric verification plate |
|
|
Multiskan Verification Plate with 8 wavelengths: 340nm, 405nm, 414nm, 450nm, 492nm, 540nm, 620nm, 690nm |
|
|
Multiskan Verification Plate with 16 wavelengths: 340nm, 405nm, 414nm, 450nm, 492nm, 540nm, 550nm, 560nm, 570nm, 580nm, 595nm, 620nm, 630nm, 650nm, 690 nm, 750nm |
|
|
Lumiwell Verification Plate |
|
Varioskan LUX 多功能微量盤分析儀可讀取多達五種不同的訊號類型,包括吸光度、螢光強度、冷光、時差性螢光以及AlphaScreen,並且提供多項配件,包括接合器、分注器和升級物件,為您的實驗提供最佳支援。請參閱Varioskan LUX 多功能微量盤分析儀之完整配件列表。
使用各種不同波長的濾片來擴展您的微量盤實驗應用。點擊以下對應的儀器名稱查看適用於機器上的濾片種類:Varioskan LUX Multimode Microplate Readers, Fluoroskan, Fluoroskan FL and Luminoskan Microplate Readers, 和 Multiskan FC Microplate Readers。
僅供研究使用,非用於臨床診斷程序。