Microplate Reader Verification Plates
Microplate Reader Verification Plates
Microplate Reader Verification Plates
Microplate Reader Verification Plates
Thermo Scientific™
Microplate Reader Verification Plates
Thermo Scientific™ Microplate Reader Verification Plates are designed to be convenient solutions for verifying and documenting the optical performance of compatible Thermo Scientific™ microplate readers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
24072800Multiskan Verification Plate (Abs), 8 Wavelengths
24072805Multiskan Verification Plate (Abs), 16 Wavelengths
2806460Lumiwell Luminometric Verification Plate (Lum)
N03394Spectrophotometric Verification Plate (Abs)
N03394M2Multifunctional Verification Plate (Abs+Fl)
Catalog number 24072800
Price (USD)
-
Product Type:
Multiskan Verification Plate (Abs), 8 Wavelengths
Thermo Scientific™ Microplate Reader Verification Plates are designed to be convenient solutions for verifying and documenting the optical performance of compatible Thermo Scientific™ microplate readers. The portfolio includes options for photometric, fluorometric, and luminometric performance verification, helping laboratories maintain confidence in measurement quality and instrument consistency over time.
Available verification plates
  • Multifunctional Verification Plate (N03394M2): Verifies photometric and fluorometric performance. Compatible with Varioskan™ LUX, Varioskan™ ALF, and Multiskan™ SkyHigh readers. Supported tests depend on the instrument model and may include photometric accuracy, precision, wavelength accuracy (verifies simultaneously fluorometric excitation wavelength accuracy) and stray light. With fluorometry, both top and bottom sensitivity and fluorescence emission wavelength accuracy can be verified.
  • Spectrophotometric Verification Plate (N03394): Verifies photometric accuracy, precision, linearity, wavelength accuracy, and stray light. Compatible with Varioskan™ LUX, Varioskan™ ALF, and Multiskan™ SkyHigh readers.
  • Multiskan Verification Plate, 8 wavelengths (24072800) or 16 wavelengths (24072805): Verifies photometric performance across 8 or 16 wavelengths. Compatible with Multiskan™ Ease (24072800) and Multiskan™ FC (24072800 or 24072805) readers.
  • Lumiwell Luminometric Verification Plate (2806460): Provides an independent Relative Light Unit reference for luminometric performance verification. Compatible with Varioskan™ LUX and Varioskan™ ALF readers.
The verification plates include certified optical reference materials appropriate for the supported detection technology. Photometric reference values are traceable to standards from the National Physical Laboratory (NPL, UK) and/or the National Institute of Standards and Technology (NIST, USA), as specified for each product. A Certificate of Calibration is supplied to support verification documentation.
The products include ready-to-use Thermo Scientific™ SkanIt™ Software sessions loading results automatically to Microsoft™ Excel™ data analysis template files to simplify measurement, result evaluation, and documentation.
Key features
  • Options for photometric, fluorometric, and luminometric verification
  • Supports routine performance verification and documentation
  • Certified reference values for reliable result evaluation
  • Designed for compatible Thermo Scientific™ microplate readers
  • Preconfigured SkanIt Software sessions and analysis files included
Includes:
  • Verification plate
  • Certificate of Calibration
  • Verification plate adapter, where required
  • USB drive with sessions, reference and data analysis template files
Protective carrying case
For Research Use Only. Not for use in diagnostic procedures.
Specifications
IncludesNPL-traceable Multiskan Verification Plate, Calibration certificate, USB with SkanIt sessions and reference files, Carrying case
Plate TypeVerification Plate
Wavelength8 Wavelengths
For Use With (Equipment)Multiskan Ease, Multiskan FC
Product TypeMultiskan Verification Plate (Abs), 8 Wavelengths
Unit SizeEach
Frequently asked questions (FAQs)
Can I calibrate the Multiskan FC Microplate Photometer (Fisherbrand accuSkan FC Filter-Based Microplate Photometer)?
The Multiskan FC Microplate Photometer (same instrument as the Fisherbrand accuSkan FC Filter-Based Microplate Photometer) is factory-calibrated and performs self-diagnostics on start-up to ensure consistent results over time. If you need to verify that the instrument is working properly, one of the options below can be utilized:
- Performance can be user-verified using Multiskan Verification Plate (Cat. No. 24072800, for 8 wavelengths or Cat. No. 24072805, for 16 wavelengths).
- The other option for verification is by purchasing our qualification service as an IQ/OQ/PQ or IPV that is required for laboratories that operate within a quality management system, comply with global standards (e.g., GLP, GCP, GMP, ISO 17025, and ISO 15189), and/or follow country-specific regulations (e.g., US FDA 21 CFR Part 11, CLIA, and CAP). Our qualification service provides documented verification that laboratory instruments are installed and operating according to the manufacturer’s performance specifications. More information about this service can be found here.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.