On this page:
Single use pipetting for precision and accuracy
Single use pipetting tools are essential in aseptic applications. Thermo Scientific Samco Transfer Pipettes allow for precise and accurate measurement and transfer of a small volume of liquid. They are ergonomic and easy to use, helping to enable efficient and controlled transfer of samples.
Key Features
Design with the end user in mind
Grips easily – no slipping
Clear, graduated markings for easy measurement
Fine-tip accuracy ensures precise liquid transfer
Package materials - reduces airborne mold spores from entering the testing area
Reliability
Inert, nontoxic polyethylene
No mold-release agents: manufactured without agents that could interfere with test results
Precision-cut tips deliver a uniform drop size
Controlled processes: a uniform bulb with uncompromised integrity`
Customizable options
Custom transfer pipettes: create pipettes specific to your requirements
Custom packaging: options for personalized design and packaging solution to meet your needs
Thermo Scientific Samco Transfer Pipettes offer reliability and precision for a variety of applications, ensuring the integrity of your samples and test results.
Select from a range of transfer pipettes to fit your application
*Exact volume is the name of this group of transfer pipettes.  These conveniently dispense a precise volume within a tolerance
Learn more about transfer pipettes
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.