Samco™ Graduated Transfer Pipettes
Actual product may vary
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Samco™ Graduated Transfer Pipettes
Thermo Scientific™

Samco™ Graduated Transfer Pipettes

Measure more than one volume accurately by using these graduated polyethylene transfer pipettes. Built-in pipette bulb eliminates need for plugging, packaging, and autoclaving.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityGraduationsPackaging TypeUnit Size
2257.7 mLNon-sterile1, 2, 3 mLBulk 500/Pk 10Pk/Cs 5000/CsCase of 5000
2123.9 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/CsCase of 5000
212PK3.9 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/CsPack of 500
212-1S3.9 mLSterile0.25, 0.50, 0.75, 1 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/CsEach
2225.8 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/CsEach
274-1S4.6 mLSterile0.5, 1, 1.5, 2 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/CsCase of 4000
212-20S3.9 mLSterile0.25, 0.50, 0.75, 1 mL20/Bag, 500/Pk., 4000/Cs.Case of 4000
222-20S5.8 mLSterile0.25, 0.50, 0.75, 1 mL20/Bag, 500/Pk., 4000/Cs.Case of 4000
225-1S7.7 mLSterile1, 2, 3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/CsCase of 4000
225-20S7.7 mLSterile1, 2, 3 mL20/Bag, 500/Pk., 4000/Cs.Case of 4000
2744.6 mLNon-sterile0.5, 1, 1.5, 2 mLBulk 500/Pk 10Pk/Cs 5000/CsCase of 5000
274PK4.6 mLNon-sterile0.5, 1, 1.5, 2 mLBulk 500/Pk 10Pk/Cs 5000/CsPack of 500
274-20S4.6 mLSterile0.5, 1, 1.5, 2 mL20/Bag, 500/Pk., 4000/Cs.Case of 4000
282TS
also known as 282
1.5 mLNon-sterile0.1 , 0.2, 0.3 mLBulk 500/Pk 8Pk/Cs 4000/CsCase of 4000
282TSPK
also known as 282
1.5 mLNon-sterile0.1 , 0.2, 0.3 mLBulk 500/Pk 8Pk/Cs 4000/CsPack of 500
282-1S1.5 mLSterile0.1, 0.2, 0.3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/CsCase of 4000
282-1SPK
also known as 282-1S
1.5 mLSterile0.1, 0.2, 0.3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/CsPack of 500
282-20S1.5 mLSterile0.1, 0.2, 0.3 mL20/Bag, 500/Pk., 4000/Cs.Case of 4000
718TS
also known as 718
5.8 mLNon-sterile0.25, 0.5, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/CsEach
728NL13.7 mLNon-sterile1, 2, 3, 4, 5 mLBulk 250/Pk 10Pk/Cs 2500/CsCase of 2500
222-1S5.8 mLSterile0.25, 0.5, 0.75, 1 mLCase of 4000
222-1SPK5.8 mLSterile0.25, 0.5, 0.75, 1 mLPack of 500
728NL-20S13.7 mLSterile1, 2, 3, 4, 5 mL20/Bag, 12 Bags/PK, 8 PK per CS, 1,920 per CSCase of 1920
Catalog number 225
Price (USD)
129.17
Online exclusive
430.00
Save 300.83
Case of 5000
Volume (Metric):
7.7 mL
Sterility:
Non-sterile
Graduations:
1, 2, 3 mL
Packaging Type:
Bulk 500/Pk 10Pk/Cs 5000/Cs
Unit Size:
Case of 5000
Price (USD)
129.17
Online exclusive
430.00
Save 300.83
Case of 5000
Accurately measure more than one volume by using Thermo Scientific™ Samco™ Graduated Transfer Pipettes. Single-unit design features a built-in pipette bulb that eliminates need for plugging, packaging, and autoclaving while preventing cross-contamination.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
Specifications
Bulb Draw (Metric)3.2 mL
DescriptionTransfer Pipette, Large Bulb, Graduated to 1mL, 2mL, 3mL
Diameter (Metric) Outer7.9 mm
Drops/mL21
GraduatedYes
Graduations1, 2, 3 mL
Packaging TypeBulk 500/Pk 10Pk/Cs 5000/Cs
DisposableSingle-use
Length (Metric)15.5 cm
MaterialLDPE
SterilityNon-sterile
TypeGraduated Transfer Pipet
Volume (Metric)7.7 mL
Unit SizeCase of 5000

Citations & References (10)

Citations & References
Abstract
Characterization of the SIM-A9 cell line as a model of activated microglia in the context of neuropathic pain.
Authors:Dave KM, Ali L, Manickam DS
Journal:PLoS One
PubMed ID:32287325
'Resident microglia of the central nervous system are being increasingly recognized as key players in diseases such as neuropathic pain. Biochemical and behavioral studies in neuropathic pain rodent models have documented compelling evidence of the critical role of ATP mediated-P2X4R-brain-derived neurotrophic factor (BDNF) signaling pathway in the initiation and maintenance ... More
Expression of the neonatal Fc-receptor in placental-fetal endothelium and in cells of the placental immune system.
Authors:Kiskova T, Mytsko Y, Schepelmann M, Helmer H, Fuchs R, Miedl H, Wadsack C, Ellinger I
Journal:Placenta
PubMed ID:30955709
'Starting from the second trimester of pregnancy, passive immunity is provided to the human fetus by transplacental transfer of maternal IgG. IgG transfer depends on the neonatal Fc receptor, FcRn. While FcRn localization in the placental syncytiotrophoblast (STB) has been demonstrated unequivocally, FcRn expression in placental-fetal endothelial cells (pFECs), which ... More
Sleep-restriction Inhibits Neurogenesis Through Decreasing the Infiltration of CD169
Authors:Yao H, Zhang Y, Xie B, Shang Y, Yuan S, Zhang J
Journal:Neuroscience
PubMed ID:32081723
'Chronic sleep-restriction (SR) is shown to be correlated with neurodevelopmental disorders. However, the effects of SR during stroke recovery on neurorepair remain unclear. In this study, mice were subjected to 60?min of cerebral ischemia followed by reperfusion. The SR protocol was accomplished by depriving mice of sleep for 20?h/day for ... More
CRISPR Interference-Based Platform for Multimodal Genetic Screens in Human iPSC-Derived Neurons.
Authors:Tian R, Gachechiladze MA, Ludwig CH, Laurie MT, Hong JY, Nathaniel D, Prabhu AV, Fernandopulle MS, Patel R, Abshari M, Ward ME, Kampmann M
Journal:Neuron
PubMed ID:31422865
CRISPR/Cas9-based functional genomics have transformed our ability to elucidate mammalian cell biology. However, most previous CRISPR-based screens were conducted in cancer cell lines rather than healthy, differentiated cells. Here, we describe a CRISPR interference (CRISPRi)-based platform for genetic screens in human neurons derived from induced pluripotent stem cells (iPSCs). We ... More
The WNT/ß-catenin signaling inhibitor XAV939 enhances the elimination of LNCaP and PC-3 prostate cancer cells by prostate cancer patient lymphocytes in vitro.
Authors:Stakheev D, Taborska P, Strizova Z, Podrazil M, Bartunkova J, Smrz D
Journal:Sci Rep
PubMed ID:30886380
Upregulated Wnt/ß-catenin signaling is associated with increased cancer cell resistance and cancer cell-elicited immunosuppression. In non-neoplastic immune cells, upregulated Wnt/ß-catenin is, however, associated with either immunosuppression or immunostimulation. Therefore, it is difficult to predict the therapeutic impact inhibitors of Wnt/ß-catenin signaling will have when combined with cancer immunotherapy. Here, we ... More