Search
View additional product information for ELISA/ELISPOT Coating Buffer Powder - FAQs (00-0044-59)
1 product FAQs found
When working with our matched pairs or uncoated ELISA kits, please refer to the user guide provided with your antibody-matched pair for kit-specific guidance. When designing your own ELISA, please utilize our ELISA Builder tool for reagent selection as well as our ELISA Development and Optimization pages for futher guidance.