Super AquaBlue ELISA Substrate, 500 mL - FAQs
Super AquaBlue ELISA Substrate, 500 mL - FAQs
View additional product information for Super AquaBlue ELISA Substrate - FAQs (00-4203-56, 00-4203-58)
2 product FAQs found
What are the differences between TMB and Super AquaBlue?
TMB is a faster developing, stronger substrate that yields greater amplification/sensitivity/background. The use of an acid stop solution produces a yellow end product read at 450nm. Super AquaBlue is a slower developing blue/green substrate particularly useful for kinetic studies.
What other reagents will I need to run an ELISA using the antibody pairs?
When working with our matched pairs or uncoated ELISA kits, please refer to the user guide provided with your antibody-matched pair for kit-specific guidance. When designing your own ELISA, please utilize our ELISA Builder tool for reagent selection as well as our ELISA Development and Optimization pages for futher guidance.