Aluminum foil, 0.13mm (0.005in) thick, 50x125mm (2.0x4.9in), 99.9995% (metals basis)
Aluminum foil, 0.13mm (0.005in) thick, 50x125mm (2.0x4.9in), 99.9995% (metals basis)
Aluminum foil, 0.13mm (0.005in) thick, 50x125mm (2.0x4.9in), 99.9995% (metals basis)
Aluminum foil, 0.13mm (0.005in) thick, 50x125mm (2.0x4.9in), 99.9995% (metals basis)
Thermo Scientific Chemicals

Aluminum foil, 0.13mm (0.005in) thick, 50x125mm (2.0x4.9in), 99.9995% (metals basis)

≈2.2g/50x125mm | CAS: 7429-90-5 | Al | 26.98 g/mol
Quantity:
4 Pk.
1 Ea.
Catalog number 000004.KI
also known as 000004-KI
Price (JPY)
-
Request A Quote
Quantity:
4 Pk.
Request bulk or custom format
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
Trace Metals≤0.0005% (≤5ppm)
FormFoil
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only