ZipChip™ Autosampler
ZipChip™ Autosampler
ZipChip™ Autosampler
ZipChip™ Autosampler
Thermo Scientific™
ZipChip™ Autosampler
Use the ZipChip™ Autosampler to prime the chips, load biological samples from vials or 96 well plates into the ZipChip Interface, and clean up between runs.
Catalog NumberFor Use With (Equipment)
00950-01-00494ZipChip Interface
Catalog number 00950-01-00494
Price (JPY)
-
For Use With (Equipment):
ZipChip Interface
ZipChip™ Autosampler is designed to work side by side with the ZipChip Interface. The ZipChip Autosampler primes the chips, loads samples from vials or 96 well plates into the ZipChip Interface, and cleans up between runs. Integrated chillers hold sensitive samples at preset temperatures while awaiting analysis. Combining rapid automation with 2-3 minute CE/ESI-MS analyses can significantly improve lab productivity and lower manual operation error rate.
Specifications
DescriptionZipChip Autosampler
Electrical Requirements110/240 V
For Use With (Equipment)ZipChip Interface
Operating Temperature4°C to Ambient -3°C (at relative humidity of 80% and ambient temperature of 25°C)<br /> Sample Tray Cooling Integrated Peltier Cooling
TypeZipChip Autosampler
Unit SizeEach