Cerium foil, 1.0mm (0.04in) thick, 99.9% (REO)
Cerium foil, 1.0mm (0.04in) thick, 99.9% (REO)
Cerium foil, 1.0mm (0.04in) thick, 99.9% (REO)
Thermo Scientific Chemicals

Cerium foil, 1.0mm (0.04in) thick, 99.9% (REO)

CAS: 7440-45-1 | Ce | 140.12 g/mol
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 010139.FI
also known as 010139-FI
Price (JPY)
-
Quantity:
50 x 50 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-45-1
IUPAC Namecerium
Molecular FormulaCe
InChI KeyGWXLDORMOJMVQZ-UHFFFAOYSA-N
SMILES[Ce]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only