Thermo Scientific Chemicals

Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010699.06
5 g, Each
Thermo Scientific Chemicals

Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010699.06
5 g, Each
Quantity
Catalog number: 010699.06
also known as 010699-06
Price (USD)
Price: 32.80
Online price: 28.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13778-31-9
IUPAC Name
copper(2+) dinitrate
Molecular Formula
CuN2O6
InChI Key
XTVVROIMIGLXTD-UHFFFAOYSA-N
SMILES
[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Metal Impurities
0.001% max.

Description

It serves as a catalyst for oxidative coupling of 2,6-dimethylphenol, resulting in an important engineering polymer material. Also used in textiles and polishing agents for other metals. It is a precursor for copper oxide, which finds use as a catalyst for many organic transformations. Menke conditions employ copper nitrate for aromatic nitrations. Claycop reagent, the hydrated copper nitrate deposited on clay, finds use in many organic reactions, for example, oxidation of dithioacetals to carbonyls and thiols to disulfides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text