Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
Quantity:
50 x 50 mm
100 x 100 mm
200 x 200 mm
Catalog number 011440.GH
also known as 011440-GH
Price (JPY)
-
Quantity:
100 x 100 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofAAg: 99.90 - 99.96% (metals basis)
Appearance (Color)Silver
FormFoil, 0.127mm (0.005in) thick, annealed.
Assay from Supplier's CofAAg + Cu: 99.94 - 100.0% (metals basis)
Assay from Supplier's CofAAl ≤ 20ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only