Cobalt(III) fluoride, 99%
Cobalt(III) fluoride, 99%
Cobalt(III) fluoride, 99%
Cobalt(III) fluoride, 99%
Thermo Scientific Chemicals

Cobalt(III) fluoride, 99%

CAS: 10026-18-3 | CoF3
Quantity:
25 g
100 g
Catalog number 011490.14
also known as 011490-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS10026-18-3
IUPAC Namecobalt(3+) trifluoride
Molecular FormulaCoF3
InChI KeyWZJQNLGQTOCWDS-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Cobalt(III) fluoride is a powerful fluorinating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only