Lead foil, 0.5mm (0.02in) thick, Puratronic, 99.99% (metalsbasis)
Lead foil, 0.5mm (0.02in) thick, Puratronic, 99.99% (metalsbasis)
Lead foil, 0.5mm (0.02in) thick, Puratronic, 99.99% (metalsbasis)
Lead foil, 0.5mm (0.02in) thick, Puratronic, 99.99% (metalsbasis)
Thermo Scientific Chemicals

Lead foil, 0.5mm (0.02in) thick, Puratronic, 99.99% (metalsbasis)

CAS: 7439-92-1 | Pb | 207.20 g/mol
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 012049.FF
also known as 012049-FF
Price (JPY)
-
Request A Quote
Quantity:
25 x 25 mm
Request bulk or custom format
Chemical Identifiers
CAS7439-92-1
IUPAC Namelead
Molecular FormulaPb
InChI KeyWABPQHHGFIMREM-UHFFFAOYSA-N
SMILES[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.998% (metals basis)
Appearance (Color)Gray
FormFoil, 0.5mm (0.02in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only