Calcium nitrate tetrahydrate, ACS, 99.0-103.0%
Calcium nitrate tetrahydrate, ACS, 99.0-103.0%
Calcium nitrate tetrahydrate, ACS, 99.0-103.0%
Thermo Scientific Chemicals

Calcium nitrate tetrahydrate, ACS, 99.0-103.0%

Manufacturing of other nitrates, fertilizers, matches | CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol
Quantity:
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012364.A3
also known as 012364-A3
Price (JPY)
-
Quantity:
2 kg
Request bulk or custom format
Chemical Identifiers
CAS13477-34-4
Molecular FormulaCaH8N2O10
InChI KeyMWGCGFYACDTFSB-UHFFFAOYSA-N
SMILESO.[Ca++].[O-][N+]([O-])=O
Molecular Weight (g/mol)236.15
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0-103.0% Ca(NO₃)₂?4H₂O
Heavy Metals(ICP-OES) 5ppm
Impurity contentK 0.005%
pHof a 5% solution 5.0-7.0 at 25?
Impurity contentBa 0.005%
View more
Calcium nitrate tetrahydrate is used as nitrogenous fertilizer and corrosive inhibitor in diesel fuels. It is utilized in plant cell for culture experiments. It is also used in cooling baths and waste water handling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only