Thermo Scientific Chemicals

Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%, Thermo Scientific Chemicals

Catalog number: 012551.22
100 g, Each
Thermo Scientific Chemicals

Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%, Thermo Scientific Chemicals

Catalog number: 012551.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 012551.22
also known as 012551-22
Price (USD)
Price: 340.00
Online price: 289.65
Your price:
Quantity
-

Chemical Identifiers

CAS
506-61-6
Total Metal Impurities
0.1% max.
Assay (unspecified)
Ag 54.2% min

Description

Silver platingPotassium silver cyanide is used in silver plating and in the manufacturing of antiseptics. It is a versatile bridging ligand and finds use for multidimensional polymer construction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text