Thermo Scientific Chemicals

Sodium dichromate dihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 012611.22
100 g, Each
Thermo Scientific Chemicals

Sodium dichromate dihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 012611.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 012611.22
also known as 012611-22
Price (USD)
Price: 32.60
Online price: 27.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7789-12-0
IUPAC Name
disodium [(oxidodioxochromio)oxy]chromiumoylolate dihydrate
Molecular Formula
Cr2H4Na2O9
InChI Key
JYDRNIYTFCBIFC-UHFFFAOYSA-N
SMILES
O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
Assay (unspecified)
99% min

Description

Sodium dichromate is used as a strong oxidizing agent in organic synthesis. It is used in metal finishing as an aid of corrosion resistance, manufacture of inorganic chromate pigments and in the preparation of colored glass and ceramic glazes. It is used as a mordant for acidic dyes to improve their color-fast properties and as a corrosion inhibitors in undercoats and primers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text