Copper silicide, 99.5% (metals basis)
Copper silicide, 99.5% (metals basis)
Copper silicide, 99.5% (metals basis)
Thermo Scientific Chemicals

Copper silicide, 99.5% (metals basis)

CAS: 12159-07-8 | Cu5Si
Quantity:
25 g
100 g
Catalog number 012844.14
also known as 012844-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS12159-07-8
IUPAC Namepentacopper silicon
Molecular FormulaCu5Si
InChI KeyJUZTWRXHHZRLED-UHFFFAOYSA-N
SMILES[Si].[Cu].[Cu].[Cu].[Cu].[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Copper silicide thin film is used for passivation of copper-based chips, where it serves to suppress diffusion and electromigration and serves as a diffusion barrier. It is involved in the direct process, the industrial route to organosilicon compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only