Thermo Scientific Chemicals

Bismuth germanium oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 013029.22
100 g, Each
Thermo Scientific Chemicals

Bismuth germanium oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 013029.22
100 g, Each
Quantity
Catalog number: 013029.22
also known as 013029-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
12233-56-6
IUPAC Name
tetrabismuth trigermanium dodecahydrate
Molecular Formula
Bi4Ge3H24O12
InChI Key
ORCSMBGZHYTXOV-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.O.O.O.[Ge].[Ge].[Ge].[Bi].[Bi].[Bi].[Bi]
Total Metal Impurities
0.0005% max.

Description

Bismuth germanium oxide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text