Lithium hydroxide monohydrate, LiOH 56.5% min
Lithium hydroxide monohydrate, LiOH 56.5% min
Lithium hydroxide monohydrate, LiOH 56.5% min
Lithium hydroxide monohydrate, LiOH 56.5% min
Thermo Scientific Chemicals

Lithium hydroxide monohydrate, LiOH 56.5% min

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol
Quantity:
50 g
500 g
2 kg
Catalog number 013409.18
also known as 013409-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)56.5% LiOH min

Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only