Praseodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Praseodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Praseodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Praseodymium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog NumberQuantity
013658.14
also known as 013658-14
25 g
Catalog number 013658.14
also known as 013658-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS13709-46-1
IUPAC Namepraseodymium(3+) trifluoride
Molecular FormulaF3Pr
InChI KeyBOTHRHRVFIZTGG-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Pr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only