Thermo Scientific Chemicals

Tantalum(V) bromide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014041.09
10 g, Each
Thermo Scientific Chemicals

Tantalum(V) bromide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014041.09
10 g, Each
Quantity
Catalog number: 014041.09
also known as 014041-09
Price (USD)
Price: 72.90
Online price: 63.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13451-11-1
IUPAC Name
tantalum(5+) pentabromide
Molecular Formula
Br5Ta
InChI Key
GCPVYIPZZUPXPB-UHFFFAOYSA-I
SMILES
[Br-].[Br-].[Br-].[Br-].[Br-].[Ta+5]
Total Metal Impurities
0.1% max.

Description

The Tantalum Bromide Cluster is used for the preparation of heavy-atom derivatives for structure determination of biological macromolecules by X-ray analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text