Ammonium sulfamate, ACS, 98% min
Ammonium sulfamate, ACS, 98% min
Ammonium sulfamate, ACS, 98% min
Thermo Scientific Chemicals

Ammonium sulfamate, ACS, 98% min

CAS: 7773-06-0 | H6N2O3S | 114.119 g/mol
Quantity:
500 g
2 kg
Catalog number 014534.36
also known as 014534-36
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7773-06-0
IUPAC Nameammonium sulfamate
Molecular FormulaH6N2O3S
InChI KeyGEHMBYLTCISYNY-UHFFFAOYSA-N
SMILES[NH4+].NS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble material0.02%
Melting Point133.0? ±2.0?
Trace Metals(as Pb) 5ppm
Assay (unspecified)98% H₆N₂O₃S min
Residue on ignition0.1%
Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. It is used as a compost accelerator, flame retardant and in industrial processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only