Thermo Scientific Chemicals

Aluminum oxide, alpha-phase, gamma-phase, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 014558.14
25 g, Each
Thermo Scientific Chemicals

Aluminum oxide, alpha-phase, gamma-phase, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 014558.14
25 g, Each
Quantity
Catalog number: 014558.14
also known as 014558-14
Price (USD)

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Total Metal Impurities
0.01% max.

Description

It is used in stainless steel coating and in microwave applications. It is used as an adsorbent, desiccating agent, grinding or blending agent, abrasive agent and catalyst, and in the manufacture of dental cements and refractories. Industrial applications include electrical and electronic products, metal Products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text