Formamide, ACS, 99.5+%
Formamide, ACS, 99.5+%
Formamide, ACS, 99.5+%
Formamide, ACS, 99.5+%
Thermo Scientific Chemicals

Formamide, ACS, 99.5+%

CAS: 75-12-7 | CH3NO | 45.04 g/mol
Quantity:
250 g
1 kg
4 x 1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014835.A1
also known as 014835-A1
Price (JPY)
-
Request A Quote
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS75-12-7
IUPAC Nameformamide
Molecular FormulaCH3NO
InChI KeyZHNUHDYFZUAESO-UHFFFAOYSA-N
SMILESNC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Assay (GC)≥99.5%
Color (APHA)10, max
CommentFreezing point 2.0-3.0?
FormLiquid
Formamide is used for cryopreservation of tissues and organs and in hybridization buffers. It acts as an RNA stabilizer in gel electrophoresis by deionizing RNA. It is used as an alternative solvent for the electrostatic self-assembly of polymer nanofilms. It is used in the preparation of brain sections of mice for BrdU immunohistochemistry and fish analysis of HeLa cells. It essentially plays a vital role in crystallization of pharmaceuticals, in soil stabilization, as a solvent in inks, as a component of liquid fertilizers, as a heat-resistant coating and in some personal care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only