Thermo Scientific Chemicals

4,4-Dimethyl-2-cyclohexen-1-one, 96%, Thermo Scientific Chemicals

1 g, Each
Thermo Scientific Chemicals

4,4-Dimethyl-2-cyclohexen-1-one, 96%, Thermo Scientific Chemicals

1 g, Each
Quantity
Catalog number: 017162.03
also known as 017162-03
Price (USD)
Price: 26.20
Your Price:
Quantity
-

Chemical Identifiers

CAS
1073-13-8
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥95.0% (non-U.S. specification)
Assay from Suppliers CofA
≥95.0% (GC, U.S. specification)
Comment
Purchased in the U.S. and in other countries

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only