Copper(II) 2-ethylhexanoate
Copper(II) 2-ethylhexanoate
Copper(II) 2-ethylhexanoate
Thermo Scientific Chemicals

Copper(II) 2-ethylhexanoate

CAS: 149-11-1 | C16H32CuO4 | 351.974 g/mol
Quantity:
10 g
50 g
Catalog number 020539.09
also known as 020539-09
Price (JPY)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS149-11-1
IUPAC Namecopper(2+) bis(2-ethylhexanoate)
Molecular FormulaC16H30CuO4
InChI KeySEKCXMNFUDONGJ-UHFFFAOYNA-L
SMILES[Cu++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder and chunks
Appearance (Color)Blue-green
Assay from Supplier's CofACopper (Cu): 16.0-19.0%
Copper(II) 2-ethylhexanoate is used as pharmaceutical intermediates. Used as drier/additive for paints, coatings and inks. Used as catalystic special applications in lubricant and grease additives, fuel combustion additives to enhance and stabilize the performance of fuels. Used in the the rubber wire coat compound and the brass-coated steel belt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only