Aluminum isopropoxide, 99.99+% (metals basis)
Aluminum isopropoxide, 99.99+% (metals basis)
Aluminum isopropoxide, 99.99+% (metals basis)
Thermo Scientific Chemicals

Aluminum isopropoxide, 99.99+% (metals basis)

CAS: 555-31-7 | C9H21AlO3 | 204.246 g/mol
Quantity:
25 g
100 g
Catalog number 022982.22
also known as 022982-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS555-31-7
IUPAC Namealuminium(3+) tris(propan-2-olate)
Molecular FormulaC9H21AlO3
InChI KeySMZOGRDCAXLAAR-UHFFFAOYSA-N
SMILES[Al+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder and/or granules
Trace Metals (by ICP)>/= 99.99% (ICP-OES)
Aluminum isopropoxide is a useful reagent in the synthesis of alkoxides, chelates and acrylates. It acts as an intermediate of pharmaceutical products like isophytol, testosterone, progesterone and ethisterone. Also used as the material of aluminate coupling agent. It is also used in alcoholysis and ester exchange reactions. In Meerwein-Ponndorf-Verley (MPV) reduction, it is used as a catalyst for the reduction of ketones and aldehydes to their corresponding alcohols. It acts as a precursor to prepare alumina.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only