Bromophenol Blue sodium salt, ACS
Bromophenol Blue sodium salt, ACS
Bromophenol Blue sodium salt, ACS
Thermo Scientific Chemicals

Bromophenol Blue sodium salt, ACS

CAS: 34725-61-6
Quantity:
5 g
25 g
100 g
Catalog number 032639.06
also known as 032639-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS34725-61-6
IUPAC Namesodium 3,5-dibromo-4-[3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular FormulaC19H9Br4NaO5S
InChI KeyKGANHHDZDYZJEM-UHFFFAOYNA-M
SMILES[Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
CommentVisual transition interval pH 3.0 (yellow) to pH 4.6 (blue)
CommentClarity of solution P.T.
Bromophenol Blue sodium salt is used as a tracking dye for protein and nucleic acid electrophoresis in polyacrylamide gels. It also used in the preparation of protein samples for western blotting analysis. It finds application in staining histones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only