Al-23 Boat;L1 (mm), 92;L2 (mm), 78;B1 (mm), 18;B2 (mm), 16;H (mm), 10;W (mm), 3;Vol, 5.7 - FAQs
View additional product information for Al-23 Boat;L1 (mm), 92;L2 (mm), 78;B1 (mm), 18;B2 (mm), 16;H (mm), 10;W (mm), 3;Vol, 5.7 - FAQs (033176.KF)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.