Al-23 Boat;L1 (mm), 92;L2 (mm), 78;B1 (mm), 18;B2 (mm), 16;H (mm), 10;W (mm), 3;Vol, 5.7
Thermo Scientific Chemicals
Al-23 Boat;L1 (mm), 92;L2 (mm), 78;B1 (mm), 18;B2 (mm), 16;H (mm), 10;W (mm), 3;Vol, 5.7
Have Questions?
Catalog NumberQuantity
033176.KF1 Pc.
Catalog number 033176.KF
Price (USD)
88.40
Each
Quantity:
1 Pc.
Request bulk or custom format
Price (USD)
88.40
Each
Specifications
Chemical Name or MaterialAl-23 Boat
Recommended StorageAmbient temperatures
TSCAU
FormBoat
Quantity1 Pc.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.