Thermo Scientific Chemicals

Osmium(III) chloride trihydrate, Premion™, 99.99% (metals basis), Os 52-56%, Thermo Scientific Chemicals

Catalog number: 033480.03
1 g, Each
Thermo Scientific Chemicals

Osmium(III) chloride trihydrate, Premion™, 99.99% (metals basis), Os 52-56%, Thermo Scientific Chemicals

Catalog number: 033480.03
1 g, Each
Quantity
Catalog number: 033480.03
Price (USD)
Price: 265.00
Online price: 225.65
Your price:
Quantity
-

Chemical Identifiers

CAS
135296-80-9
IUPAC Name
trichloroosmium trihydrate
Molecular Formula
Cl3H6O3Os
InChI Key
UACQLNPCDXDCID-UHFFFAOYSA-K
SMILES
O.O.O.Cl[Os](Cl)Cl
Total Metal Impurities
0.01% max.

Description

Osmium(III) chloride trihydrate is used as a reagent for preparation of dichlorodihydridoosmium complexes. It is also useful as starting materials and an efficient alkane oxidation catalyst in the presence of pyridine and hydrogen oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text