Interference Check Standard AB-1(ICS-AB1) Solution, Specpure™
Thermo Scientific Chemicals

Interference Check Standard AB-1(ICS-AB1) Solution, Specpure™

 
Catalog NumberQuantity
035421.AE
also known as 035421-AE
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035421.AE
also known as 035421-AE
Price (JPY)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialInterference Check Standard AB-1(ICS-AB1) Solution
DOT InformationHazard Class: 8; Packaging Group: II
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only