Thermo Scientific Chemicals

Sodium aluminum oxide, tech., Thermo Scientific Chemicals

Catalog number: 035453.22
100 g, Each
Thermo Scientific Chemicals

Sodium aluminum oxide, tech., Thermo Scientific Chemicals

Catalog number: 035453.22
100 g, Each
Quantity
Catalog number: 035453.22
also known as 035453-22
Price (USD)
Price: 64.90
Online price: 55.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1302-42-7
IUPAC Name
sodium dioxoalumanuide
Molecular Formula
AlNaO2
InChI Key
IYJYQHRNMMNLRH-UHFFFAOYSA-N
SMILES
[Na+].O=[Al-]=O
Insoluble material
1% max

Description

Sodium aluminum oxide is used in the isomerization of alkene and isomerization of amines. It is also used in water treatment as an adjunct and as a coagulant aid to flocculation. It is involved in the removal of silica and phosphates from waste water. It is employed to accelerate concrete solidification. It finds application in the paper industry and fire brick production. It acts as an intermediate in the production of zeolites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text