Sodium hydroxide, 5.0N Standardized Solution
Sodium hydroxide, 5.0N Standardized Solution
Sodium hydroxide, 5.0N Standardized Solution
Thermo Scientific Chemicals

Sodium hydroxide, 5.0N Standardized Solution

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Quantity:
1 L
4 L
Catalog number 035633.K7
also known as 035633-K7
Price (JPY)
-
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration5.0N NaOH
Sodium hydroxide, 5.0N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only