Ammonium carbonate, ACS, NH{3} 30.0+%
Ammonium carbonate, ACS, NH{3} 30.0+%
Ammonium carbonate, ACS, NH{3} 30.0+%
Ammonium carbonate, ACS, NH{3} 30.0+%
Thermo Scientific Chemicals

Ammonium carbonate, ACS, NH{3} 30.0+%

In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue | CAS: 506-87-6 | CH8N2O3 | 96.086 g/mol
Quantity:
500 g
2 kg
Catalog number 036229.A3
also known as 036229-A3
Price (JPY)
-
Request A Quote
Quantity:
2 kg
Request bulk or custom format
Chemical Identifiers
CAS506-87-6
IUPAC Namecarbonic acid diamine
Molecular FormulaCH8N2O3
InChI KeyPRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILESN.N.OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)30.0% NH₃ min
Heavy Metals(ICP-OES) 5ppm
Impurity contentCl 5ppm
Impurity contentFe 5ppm
Impurity contentNonvolatile matter 0.01%
View more
In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compoundsAmmonium carbonate is widely utilized as a leavening agent in lebkuchen, cookies and flat biscuits. It finds an important application as an emetic and an active ingredient

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds
RUO – Research Use Only