Potassium chlorate, ACS, 99.0% min
Potassium chlorate, ACS, 99.0% min
Potassium chlorate, ACS, 99.0% min
Thermo Scientific Chemicals

Potassium chlorate, ACS, 99.0% min

CAS: 3811-04-9 | ClKO3 | 122.55 g/mol
Quantity:
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036494.36
also known as 036494-36
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS3811-04-9
IUPAC Namepotassium chlorate
Molecular FormulaClKO3
InChI KeyVKJKEPKFPUWCAS-UHFFFAOYSA-M
SMILES[K+].[O-][Cl](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metals(as Pb) 5ppm
Assay (unspecified)99.0% KCLO₃ min
Impurity contentCl 0.001%
Impurity contentMg 0.002%
Impurity contentCa 0.002%
View more
It is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only