Thermo Scientific Chemicals

Acrylamide, 99.9%, Thermo Scientific Chemicals

Catalog number: 036545.22
100 g, Each
Thermo Scientific Chemicals

Acrylamide, 99.9%, Thermo Scientific Chemicals

Catalog number: 036545.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 036545.22
also known as 036545-22
Price (USD)
Price: 104.00
Online price: 88.65
Your price:
Quantity
-

Chemical Identifiers

CAS
79-06-1
IUPAC Name
prop-2-enamide
Molecular Formula
C3H5NO
InChI Key
HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES
NC(=O)C=C
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥99.9%
Conductivity
≤50 µmhos
DNase
None detected

Description

Acrylamide is primarily used in research laboratories for electrophoresis, chromatography, electron microscopy and in the sewage and wastewater treatment industries. It is used in the production of polymers mainly in the pulp and paper industry and textile treatment industry. The largest use for acrylamide is as an intermediate in the production of organic chemicals and in the synthesis of polyacrylamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text