Diamond powder, natural, 40-60 micron
Thermo Scientific Chemicals

Diamond powder, natural, 40-60 micron

Diamond powder is generally used as an abrasive to grind and polish hard surfaces
Quantity:
1 g
5 g
Catalog number 038984.06
also known as 038984-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
Chemical Name or MaterialDiamond powder
EINECS Number231-953-2
MDL NumberMFCD00211867
Name NoteNatural
View more
Diamond powder is generally used as an abrasive to grind and polish hard surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only