Thermo Scientific Chemicals

Cerium(IV) oxide, 96% (REO), Thermo Scientific Chemicals

Catalog number: 040581.30
250 g, Each
Thermo Scientific Chemicals

Cerium(IV) oxide, 96% (REO), Thermo Scientific Chemicals

Catalog number: 040581.30
250 g, Each
Quantity
Catalog number: 040581.30
also known as 040581-30
Price (USD)
Price: 145.00
Online price: 125.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1306-38-3
IUPAC Name
dioxocerium
Molecular Formula
CeO2
InChI Key
CETPSERCERDGAM-UHFFFAOYSA-N
SMILES
O=[Ce]=O
Total Rare Earth Oxide Impurities
4% max.

Description

Cerium(IV) oxide is used in polishing and decolorizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Polishing and decolorizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text