Polypropylene glycol 400
Polypropylene glycol 400
Polypropylene glycol 400
Thermo Scientific Chemicals

Polypropylene glycol 400

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol
Quantity:
50 g
250 g
Catalog number 040811.30
also known as 040811-30
Price (JPY)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS25322-69-4
Molecular Formula(C3H6O)n
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES[H]OC(C)CO
MDL NumberMFCD00084446
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Formviscous liquid
It is used in cosmetic and pharmaceutical preparations and in the manufacture of emulsifying or wetting agents and lubricants,suppress foaming in industrial processes and for making polyurethane resins, hydraulic fluids, and various other materials. It acts as swelling agent for the preparation and characterization of organo-modified SBA-15. Polypropylene glycol and tetraethoxysilane precursors play an important role for the preparation of superhydrophobic silica-based surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only