Tungsten wire, gold plated, 0.05mm (0.002in) dia, W 99.95% (metals basis), Au 99.99%
Thermo Scientific Chemicals

Tungsten wire, gold plated, 0.05mm (0.002in) dia, W 99.95% (metals basis), Au 99.99%

Quantity:
1 m
5 m
10 m
Catalog number 041039.G5
also known as 041039-G5
Price (JPY)
-
Quantity:
5 m
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
Chemical Name or MaterialTungsten wire, gold plated
Concentration or Composition (by Analyte or Components)Au 99.99%
MDL NumberMFCD00011461
Name Note0.05mm (0.002 in.) dia.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only