Sodium tellurite(IV), 99.5% (metals basis)
Sodium tellurite(IV), 99.5% (metals basis)
Sodium tellurite(IV), 99.5% (metals basis)
Thermo Scientific Chemicals

Sodium tellurite(IV), 99.5% (metals basis)

CAS: 10102-20-2 | H10Na2O9Te | 327.65 g/mol
Quantity:
10 g
50 g
Catalog number 041778.18
also known as 041778-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS10102-20-2
IUPAC Namedisodium pentahydrate tellurate
Molecular FormulaH10Na2O9Te
InChI KeyOVBVNEXDNNHXIU-UHFFFAOYSA-L
SMILESO.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Sodium tellurite(IV) acts as an intermediate used in the extraction of the tellurium. It is used to make corrosion-resistant electroplated nickel. It is also used to add a bluish coating to iron, steel and aluminum. It serves as a growth medium for bacteria in microbiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only