Thermo Scientific Chemicals
Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number: 041802.GH
100 x 100 mm, Each
Thermo Scientific Chemicals
Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number: 041802.GH
100 x 100 mm, Each
Quantity
Catalog number: 041802.GH
also known as 041802-GH
Price (USD)
Price: 1,642.00
Your price:
Quantity
-
Description
Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis) is used in the fabrication of micro electrodes for electrical stimulation of nervous tissue and electrophysiological recordings.