Dichloromethane, anhydrous, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, stab. with amylene
Dichloromethane, anhydrous, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, stab. with amylene
Dichloromethane, anhydrous, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, stab. with amylene
Thermo Scientific Chemicals

Dichloromethane, anhydrous, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, stab. with amylene

Solvent For Anhydrous Reactions, Water <0.01% | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041835.AK
also known as 041835-AK
Price (USD)
-
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation3 ppm. Max
Assay (GC)99.7% min
Water Content (Karl Fischer Titration)0.01% max

Solvent for anhydrous reactions, water <0.01%.Dichloromethane is employed in pharmaceutical applications. Its utilization is as a solvent in manufacturing of fine chemicals, drugs, drug intermediates and pharmaceuticals, and also in food engineering. It is the solvent of choice for caffeine extraction. It is employed as a solvent and paint remover, and as a blowing agent for polyurethane foams. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only