Thermo Scientific Chemicals

Arsenic(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 041980.06
5 g, Each
Thermo Scientific Chemicals

Arsenic(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 041980.06
5 g, Each
Quantity
Catalog number: 041980.06
also known as 041980-06
Price (USD)

Chemical Identifiers

CAS
1303-33-9
IUPAC Name
diarsenic(3+) trisulfanediide
Molecular Formula
As2S3
InChI Key
CUGMJFZCCDSABL-UHFFFAOYSA-N
SMILES
[S--].[S--].[S--].[As+3].[As+3]
Total Metal Impurities
0.001% max.

Description

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text