Ammonium iron(III) oxalate hydrate, Thermo Scientific Chemicals
Ammonium iron(III) oxalate hydrate, Thermo Scientific Chemicals
Ammonium iron(III) oxalate hydrate, Thermo Scientific Chemicals
Ammonium iron(III) oxalate hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium iron(III) oxalate hydrate, Thermo Scientific Chemicals

In photography, blueprints
Quantity:
50 g
250 g
Catalog number 042112.18
also known as 042112-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Specifications
CAS13268-42-3
Chemical Name or MaterialAmmonium iron(III) oxalate hydrate
DOT InformationHazard Class: 8; Packaging Group: III
Formula Weight374.02 (Anhydrous)
Health Hazard 1H302-H312-H314-H335
View more
In photography, blueprints

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only