Thermo Scientific Chemicals

Potassium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 042217.14
25 g, Each
Thermo Scientific Chemicals

Potassium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 042217.14
25 g, Each
Quantity
Catalog number: 042217.14
also known as 042217-14
Price (USD)

Chemical Identifiers

CAS
7789-23-3
IUPAC Name
potassium fluoride
Molecular Formula
FK
InChI Key
NROKBHXJSPEDAR-UHFFFAOYSA-M
SMILES
[F-].[K+]
Total Metal Impurities
0.1% max.

Description

A versatile fluoride ion source used in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides. This ion source is used recently in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text