Buffer solution, pH 11.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable
Buffer solution, pH 11.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable
Buffer solution, pH 11.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable
Thermo Scientific Chemicals

Buffer solution, pH 11.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable

CAS: 1336-21-6 | H5NO | 35.05 g/mol
Quantity:
500 mL
1 L
4 L
Catalog number 042424.K7
also known as 042424-K7
Price (JPY)
-
Quantity:
4 L
Request bulk or custom format
Specifications
CAS1336-21-6
ChEBICHEBI:18219
Chemical Name or MaterialBuffer solution
ColorColorless
IUPAC Nameazanium;hydroxide
View more
Buffer solution (pH 11) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only