Lithium, Oil based standard solution, Specpure™ Li 5000μg/g
Lithium, Oil based standard solution, Specpure™ Li 5000μg/g
Lithium, Oil based standard solution, Specpure™ Li 5000μg/g
Thermo Scientific Chemicals

Lithium, Oil based standard solution, Specpure™ Li 5000μg/g

 
Catalog NumberQuantity
042442.18
also known as 042442-18
50 g
Catalog number 042442.18
also known as 042442-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium, Oil based standard solution
MDL NumberMFCD00134051
Name NoteLi 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only